Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Thrombospondin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7860
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7860 Supplier R&D Systems Supplier No. AF7860
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Thrombospondin-4 Polyclonal specifically detects Thrombospondin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Thrombospondin-4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z1T2
Gene Alias THBS4, thrombospondin 4, thrombospondin-4, Thrombospondin4, TSP4, TSP-4
Gene Symbols THBS4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Thrombospondin-4 Gln27-Asn963 (predicted) Accession # Q9Z1T2
Purification Method Immunogen affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7060
Test Specificity Detects mouse Thrombospondin-4 in direct ELISAs and Western blots. In direct ELISAs, aproximately 10% cross-reactivity with recombinant human Thrombospondin-4 is observed, and less than 1% cross-reactivity with recombinant mouse Thrombospondin-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.