Learn More
Novus Biologicals™ Mouse Thyroxine Autoantibody ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Sensitivity | 1.88 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Mouse T4-Ab in samples. No Significant cross-reactivity or interference between Mouse T4-Ab and analogues was observed. This kit does not detect a specific autoantibody isotype. |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Measuring Range | 3.13 to 200 ng/mL |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Thyroxine Autoantibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.