Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIGIT Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $573.00
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | 2190A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB72671SP
![]() |
R&D Systems
MAB72671SP |
25 μg |
Each for $227.50
|
|
|||||
MAB72671100
![]() |
R&D Systems
MAB72671100 |
100 μg |
Each for $573.00
|
|
|||||
Description
TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| TIGIT | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P86176 | |
| 201633 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2190A | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIGIT Met1-Thr143 Accession # P86176 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TIGIT in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title