Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TIGIT Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7267100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7267100 100 μg
MAB7267SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7267100 Supplier R&D Systems Supplier No. MAB7267100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen TIGIT
Applications Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 826118
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction 40-240 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P86176
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TIGIT Met1-Thr143 Accession # P86176
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Test Specificity Detects mouse TIGIT in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.