Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TIGIT APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7267A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7267A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7267A Supplier R&D Systems Supplier No. FAB7267A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

TIGIT Polyclonal specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TIGIT
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. NP_001139797
Gene Symbols Tigit
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TIGIT Gly26-Thr143 Accession # NP_001139797
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Test Specificity Detects mouse TIGIT in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human TIGIT is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.