Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-1/KIM-1/HAVCR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 22 publications
Supplier: R&D Systems AF1817SP
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot.Specifications
TIM-1/KIM-1/HAVCR | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
26762 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.25 ug/mL | |
NP_001160104 | |
HAVCR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104 | |
25 μg | |
Primary | |
Detects mouse TIM-1/KIM-1/HAVCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human TIM-1, recombinant mouse (rm) TIM-2, rmTIM-3, rmTIM-4, rmTIM-6, and rmTIM-7 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction