Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-1/KIM-1/HAVCR Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 11 publications
$214.00 - $2020.00
Specifications
Antigen | TIM-1/KIM-1/HAVCR |
---|---|
Clone | 222414 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1817SP
![]() |
R&D Systems
MAB1817SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1817100
![]() |
R&D Systems
MAB1817100 |
100 μg |
Each for $573.00
|
|
|||||
MAB1817500
![]() |
R&D Systems
MAB1817500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
TIM-1/KIM-1/HAVCR | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
NP_001160104 | |
26762 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TIM-1/KIM-1/HAVCR in ELISAs and Western blots. This antibody does not cross-react with recombinant human (rh) TIM-1, rmTIM-2, or rmTIM-3. |
222414 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
HAVCR1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title