Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-1/KIM-1/HAVCR Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems BAF1817
This item is not returnable.
View return policy
Description
TIM-1/KIM-1/HAVCR Polyclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
TIM-1/KIM-1/HAVCR | |
Polyclonal | |
Biotin | |
NP_001160104 | |
HAVCR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-1 (R&D Systems, Catalog # 1817-TM) Tyr22-Thr212 Accession # NP_001160104 | |
50 μg | |
Primary | |
Detects mouse TIM-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human TIM-1, rhTIM-3, rhTIM-4, recombinant mouse (rm) TIM-4, rmTIM-3 and recombinant rat KIM-1 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
26762 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction