Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | TIM-3 |
---|---|
Clone | 215015 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15291SP
![]() |
R&D Systems
MAB15291SP |
25 μg |
Each for $200.50
|
|
|||||
MAB15291
![]() |
R&D Systems
MAB15291 |
500 μg |
Each for $667.00
|
|
|||||
Description
TIM-3 Monoclonal specifically detects TIM-3 in Mouse samples. It is validated for Western Blot.Specifications
TIM-3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q8VIM0 | |
84868 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-3 Arg20-Ala193 Accession # Q8VIM0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TIM-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) TIM-1, recombinant mouse (rm) TIM-1, rmTIM-2, or rhTIM-3. |
215015 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
HAVCR2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title