Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1529
This item is not returnable.
View return policy
Description
TIM-3 Polyclonal specifically detects TIM-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
TIM-3 | |
Polyclonal | |
Unconjugated | |
AAL65156 | |
HAVCR2 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-3 Leu22-Arg191 Accession # AAL65156 | |
100 μg | |
Primary | |
Detects mouse TIM-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human TIM-3 is observed and less than 5% cross-reactivity with recombinant mouse (rm) TIM-1, rmTIM-2, rmTIM-4, rmTIM-5, rmTIM-6, and rmTIM-7 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 | |
Goat | |
Affinity Purified | |
RUO | |
84868 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction