Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | TIM-4 |
---|---|
Clone | 1018144 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2826SP
![]() |
R&D Systems
MAB2826SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2826100
![]() |
R&D Systems
MAB2826100 |
100 μg |
Each for $432.50
|
|
|||||
Description
TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
TIM-4 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
Mouse myeloma cell line, NS0-derived mouse TIM-4, Ala22-Thr279, Accession # Q6U7R4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1018144 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
Q6U7R4 | |
91937 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title