Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3374SP
This item is not returnable.
View return policy
Description
TIM-5 Monoclonal specifically detects TIM-5 in Mouse samples. It is validated for Western Blot.Specifications
TIM-5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_001156829 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
210535 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
339319 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Gm12169 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 Accession # NP_001156829 | |
25 μg | |
Primary | |
Detects mouse TIM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TIM-1, -2, -3, -4, or -6 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction