Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIM-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
$164.60 - $461.52
Specifications
| Antigen | TIM-5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TIM-5 Polyclonal specifically detects TIM-5 in Mouse samples. It is validated for Western Blot.Specifications
| TIM-5 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| 210535 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-5 Phe22-Gly161 Accession # NP_001156829 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse TIM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse (rm) TIM-6 is observed and less than 1% cross-reactivity with rmTIM-2, rmTIM-3, and rmTIM-4 is observed. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001156829 | |
| Gm12169 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title