Learn More
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA Development Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Mouse |
| Conjugate | Biotin |
| Binding Target | TIMP-2 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7077 |
| Product Line | R&D Systems |
| Synonym | CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP2, TIMP-2, Tissue inhibitor of metalloproteinases 2 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
