Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TL1A/TNFSF15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1896SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1896SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1896SP Supplier R&D Systems Supplier No. MAB1896SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Mouse samples. It is validated for ELISA, Neutralization.

Specifications

Antigen TL1A/TNFSF15
Applications Neutralization
Classification Monoclonal
Clone 820446
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.8-4 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5UBV8
Gene Alias MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1
Gene Symbols TNFSF15
Host Species Rat
Immunogen E. coli-derived recombinant mouse TL1A/TNFSF15 Ile76-Leu252 Accession # Q5UBV8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9966
Test Specificity Detects mouse TL1A/TNFSF15 in ELISA.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.