Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TL1A/TNFSF15 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

$200.50 - $432.50

Specifications

Antigen TL1A/TNFSF15
Clone 820422
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB18961SP
GREENER_CHOICE
View Documents
R&D Systems
MAB18961SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB18961100
GREENER_CHOICE
View Documents
R&D Systems
MAB18961100
100 μg
Each for $432.50
Only null left
Add to Cart
 
Description

Description

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Mouse samples. It is validated for ELISA.
Specifications

Specifications

TL1A/TNFSF15
ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1
E. coli-derived recombinant mouse TL1A/TNFSF15, Ile76-Leu252, Accession # Q5UBV8
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse TL1A/TNFSF15 in direct ELISAs.
820422
ELISA
Unconjugated
Rat
Mouse
Q5UBV8
9966
IgG2c
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.