Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1475
This item is not returnable.
View return policy
Description
TLR1 Polyclonal specifically detects TLR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
TLR1 | |
Polyclonal | |
Unconjugated | |
Q9EPQ1 | |
TLR1 | |
Mouse myeloma cell line NS0-derived recombinant mouse TLR1 Ser25-Asp581 Accession # Q9EPQ1 | |
100 μg | |
Primary | |
Detects mouse TLR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) TLR6 is observed, 5% cross-reactivity with recombinant human (rh) TLR1 and rmTLR2 is observed, and less than 2% cross-reactivity with rhTLR3 and rhTLR4 is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
7096 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction