Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR11 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB7640
This item is not returnable.
View return policy
Description
TLR11 Monoclonal specifically detects TLR11 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
TLR11 | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q6R5P0 | |
TLR11 | |
E. coli-derived recombinant mouse TLR11 Thr209-Gln325 Accession # Q6R5P0 | |
100 μg | |
Primary | |
Detects mouse TLR11 in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
786404 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Gm287, hgnc31753, Toll like receptor 11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
442887 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction