Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR12 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC8086G100UG
This item is not returnable.
View return policy
Description
TLR12 Monoclonal specifically detects TLR12 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
TLR12 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Gm1365, toll-like receptor 12 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR12 Thr22-Lys709 Accession # Q6QNU9 | |
RUO | |
100131451 | |
Mouse | |
IgG |
Flow Cytometry | |
1229C | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6QNU9 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse TLR12 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction