Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
$200.50 - $667.00
Specifications
Antigen | TLR2 |
---|---|
Clone | 203325 |
Concentration | LYOPH |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
TLR2 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
Q9QUN7 | |
7097 | |
Mouse myeloma cell line NS0-derived recombinant mouse TLR2 Gln25-Leu590 Accession # Q9QUN7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TLR2 in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant mouse (rm) TLR1, recombinant human (rh) TLR2, rhTLR3, rhTLR4, or rmTLR6. |
203325 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD282, CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
TLR2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title