Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3005
This item is not returnable.
View return policy
Description
TLR3 Polyclonal specifically detects TLR3 in Mouse samples. It is validated for Western Blot.Specifications
TLR3 | |
Polyclonal | |
Unconjugated | |
Q99MB1 | |
TLR3 | |
Mouse myeloma cell line NS0-derived recombinant mouse TLR3 (R&D Systems, Catalog # 3005-TR) Cys29-Leu705 Accession # Q99MB1 | |
100 μg | |
Primary | |
Detects mouse TLR3 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human TLR3 is observed, 5% cross-reactivity with recombinant mouse (rm) TLR4 is observed, and less than 1% cross-reactivity with rmTLR2 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
CD283, CD283 antigen, toll-like receptor 3 | |
Goat | |
Affinity Purified | |
RUO | |
7098 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction