Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR4 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB27591G100UG
This item is not returnable.
View return policy
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry.Specifications
TLR4 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Mouse myeloma cell line NS0-derived recombinant mouse TLR4 Asn26-Lys629 Accession # Q9QUK6 | |
RUO | |
7099 | |
Mouse | |
IgG |
Flow Cytometry | |
1203B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9QUK6 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse TLR4 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction