Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TLR9 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB7960S100UG
This item is not returnable.
View return policy
Description
TLR9 Monoclonal specifically detects TLR9 in Mouse samples. It is validated for Flow Cytometry.Specifications
TLR9 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD289, CD289 antigen, toll-like receptor 9 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9 Leu26-Asp818 Accession # AAK29625 | |
RUO | |
54106 | |
Mouse | |
IgG |
Flow Cytometry | |
1138D | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAK29625 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse TLR-9 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction