Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TLR9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7960SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7960SP 25 μg
MAB7960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7960SP Supplier R&D Systems Supplier No. MAB7960SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TLR9 Monoclonal specifically detects TLR9 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.

Specifications

Antigen TLR9
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1138D
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 0.5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. AAK29625
Gene Alias CD289, CD289 antigen, toll-like receptor 9
Gene Symbols Tlr9
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TLR9 Leu26-Asp818 Accession # AAK29625
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54106
Test Specificity Detects mouse TLR-9 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.