Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TNF-alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
$241.00 - $549.50
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | 653812R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4102SP
![]() |
R&D Systems
MAB4102SP |
25 μg |
Each for $241.00
|
|
|||||
MAB4102100
![]() |
R&D Systems
MAB4102100 |
100 μg |
Each for $549.50
|
|
|||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Mouse samples. It is validated for ELISA.Specifications
| TNF-alpha | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| P06804 | |
| 7124 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 653812R | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| E. coli-derived recombinant mouse TNF-alpha Leu80-Leu235, with an N-terminal Met & Ser84-Leu235 Accession # P06804 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects mouse TNF-alpha in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title