Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TNF-alpha Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 27 publications
Supplier: R&D Systems BAF410
This item is not returnable.
View return policy
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
| TNF-alpha | |
| Polyclonal | |
| Biotin | |
| P06804 | |
| TNF | |
| E. coli-derived recombinant mouse TNF-alpha (R&D Systems, Catalog # 410-MT) Leu80-Leu235 Accession # P06804 | |
| 50 μg | |
| Primary | |
| Detects mouse TNF-alpha in ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7124 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction