Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse TNF-alpha Quantikine HS ELISA Kit
SDP

Catalog No. MHSTA50
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHSTA50 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MHSTA50 Supplier R&D Systems™ Supplier No. MHSTA50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

E. coli-expressed recombinant mouse TNF-alpha and antibodies raised against the recombinant factor

Mouse TNF-alpha Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse TNF-alpha Quantikine HS ELISA Kit quantifies mouse TNF-alpha in serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Description APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, tnfa, TNF-a, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Product Type ELISA Kit
Quantity 1 Kit
Species Mouse
Assay Sensitivity 0.295 pg/mL
Assay Range 0.78 to 50 pg/mL
Cross Reactivity <0.5% cross-reactivity observed with available related molecules, <50% cross-species reactivity observed with species tested
For Use With (Application) ELISA
Gene ID (Entrez) 7124
Sample Type Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Sample Volume 50 μL (Serum), 50 μL (Heparin plasma), 50 μL (EDTA plasma)
Test Time 4 hr.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Kit Contents E. coli-expressed recombinant mouse TNF-alpha and antibodies raised against the recombinant factor
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.