Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TNF RI/TNFRSF1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB430500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB430500 500 μg
MAB430SP 25 μg
MAB430100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB430500 Supplier R&D Systems Supplier No. MAB430500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hamster Monoclonal Antibody has been used in 9 publications

TNF RI/TNFRSF1A Monoclonal specifically detects TNF RI/TNFRSF1A in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.

Specifications

Antigen TNF RI/TNFRSF1A
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 55R170
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation, Neutralization 5-20 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNFRI, TNF-RI, TNF-R-I, TNFR-I, TNFRSF1A, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Gene Symbols TNFRSF1A
Host Species Hamster
Immunogen Recombinant mouse TNF RI/TNFRSF1A Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7132
Test Specificity Detects mouse TNF RI/TNFRSF1A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, recombinant mouse (rm) CD27, rmCD30, rhCD40, rmCD40, rhDR3, rhDR6, rmEDAR, rhFas, rmFas, rhEDAR, rhGITR, rmGITR, rhHVEM, rhLT beta R, rmLT beta R, rhNGF R, rhOPG, rmOPG, rhRANK, rmRANK, rhTAJ, rhTNF RI, rhTNF RII or rmTNF RII is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.