Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TNF RII/TNFRSF1B Alexa Fluor 405-conjugated Antibody, R&D Systems™

Hamster Monoclonal Antibody
Supplier: R&D Systems FAB426V100UG
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal specifically detects TNF RII/TNFRSF1B in Mouse samples. It is validated for Flow Cytometry.Specifications
TNF RII/TNFRSF1B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNFRII, TNF-RII, TNF-R-II, TNFR-II, TNFRSF1B, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II | |
Hamster | |
100 μg | |
Primary | |
Detects mouse TNF RII/TNFRSF1B in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) TNF RII, recombinant mouse (rm) TNF RI, rhTNF RI, rm4-1BB, or rmOPG is observed. In Western blots, approximately 25% cross-reactivity with rhDR3 and rhTNF RII is observed and no cross-reactivity with rm4-1BB, rmCD27, rmCD30, rmCD40, rmFas, rmGITR, rhHVEM, rhNGF R, rmOPG, rmRANK, rmTNF RI, rmTROY, or rmEDAR is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
TR7554 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
TNFRSF1B | |
E. coli-derived recombinant mouse TNF RII/TNFRSF1B Extracellular domain | |
RUO | |
7133 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction