Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TNFRH3/TNFRSF26 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5330U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5330U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5330U100 Supplier R&D Systems Supplier No. FAB5330U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TNFRH3/TNFRSF26 Monoclonal specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TNFRH3/TNFRSF26
Applications Flow Cytometry
Classification Monoclonal
Clone 509027
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P83626
Gene Symbols Tnfrsf26
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26 Ser20-Lys162 Accession # P83626
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Detects mouse TNFRH3/TNFRSF26 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Fas, rmTNF RI, and rmTNF RII is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.