Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TNFRH3/TNFRSF26 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5330 Supplier R&D Systems Supplier No. MAB5330
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TNFRH3/TNFRSF26 Monoclonal specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen TNFRH3/TNFRSF26
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 509027
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P83626
Gene Symbols Tnfrsf26
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26 Ser20-Lys162 Accession # P83626
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Detects mouse TNFRH3/TNFRSF26 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) Fas, rmTNF RI, and rmTNF RII is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.