Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TORC3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6840
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6840 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6840 Supplier R&D Systems Supplier No. MAB6840
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TORC3/CRTC3 Monoclonal specifically detects TORC3/CRTC3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TORC3/CRTC3
Applications Western Blot
Classification Monoclonal
Clone 643729
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q91X84
Gene Alias CREB regulated transcription coactivator 3, CRTC3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3
Gene Symbols Crtc3
Host Species Rat
Immunogen E. coli-derived recombinant mouse TORC3 Leu153-Pro275 Accession # Q91X84
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64784
Test Specificity Detects mouse TORC3 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human TORC3 and no cross-reactivity with recombinant mouse (rm) TORC1 or rmTORC2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.