Learn More
Description
- This kit recognizes Mouse ACP5 in samples.
- No significant cross-reactivity or interference between Mouse ACP5 and analogues was observed.
- Assay Type: Sandwich ELISA
- Standard Curve Range: 0.78 - 50 ng/mL
- Sensitivity: 0.47 ng/mL
- Inter-Assay: 5.02%
- Intra-Assay: 5.26%
- Spike Recovery: 89-108%
- Sample Volume: 100 μL
- Components:
- Concentrated Wash Buffer (25×)
- Concentrated Biotinylated Detection Ab (100×)
- Concentrated HRP Conjugate (100×)
- Certificate of Analysis
- Biotinylated Detection Ab Diluent
- HRP Conjugate Diluent
- Plate Sealer
- Micro ELISA Plate(Dismountable)
- Product Description
- Reference Standard
- Stop Solution
- Reference Standard & Sample Diluent
- Substrate Reagent
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Gene Alias | HPAP, TRACP5a, TRACP5b, TRAP, TrATPase |
| Gene ID (Entrez) | 54 |
| Gene Symbol | ACP5 |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
