Learn More
Mouse TRAIL R2/TNFRSF10B Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 3 publications
Supplier:  R&D Systems AF721SP
Catalog No. AF721SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
TRAILR2/TNFRSF10B Polyclonal specifically detects TRAILR2/TNFRSF10B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.
Specifications
Specifications
TRAILR2/TNFRSF10B
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD262, CD262 antigen, death domain containing receptor for TRAIL/Apo-2L, Death receptor 5, DR5, DR5TRICK2B, Fas-like protein, KILLER/DR5, KILLERapoptosis inducing protein TRICK2A/2B, TNF-related apoptosis-inducing ligand receptor 2, TNFRSF10B, TRAIL receptor 2, TRAILR2, TRAIL-R2apoptosis inducing receptor TRAIL-R2, TRAILR2cytotoxic TRAIL receptor-2, TRICK2, TRICK2A, TRICKB, tumor necrosis factor receptor superfamily member 10B, tumor necrosis factor receptor superfamily, member 10b, tumor necrosis factor receptor-like protein ZTNFR9, ZTNFR9
Goat
Affinity Purified
RUO
8795
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Flow Cytometry, Neutralization, CyTOF
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.4-1.2 ug/mL, CyTOF-ready
Q6GSD9
TNFRSF10B
Mouse myeloma cell line NS0-derived recombinant mouse TRAIL R2/TNFRSF10B Asn53-Ser177 Accession # Q6GSD9
25 μg
Primary
Detects mouse TRAIL R2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) TRAIL R2 is observed.
Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only