Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TRAM/TICAM2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4348
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4348 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4348 Supplier R&D Systems Supplier No. MAB4348
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TRAM/TICAM2 Monoclonal specifically detects TRAM/TICAM2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TRAM/TICAM2
Applications Western Blot
Classification Monoclonal
Clone 757706
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8BJQ4
Gene Alias MGC129876, MGC129877, Putative NF-kappa-B-activating protein 502, TICAM2, TICAM-2TIR domain-containing adapter molecule 2, TIRAP3, TIRP, TIRPcytoplasmic adaptor, toll/interleukin-1 receptor (TIR) domain-containing adapter protein, Toll/interleukin-1 receptor domain-containing protein, toll-like receptor adaptor molecule 2, Toll-like receptor adaptor protein 3, TRAMNF-kappa-B-activating protein 502, Trif, TRIF-related adapter molecule, TRIF-related adaptor molecule
Gene Symbols TICAM2
Host Species Rat
Immunogen E. coli-derived recombinant mouse TRAM/TICAM2 Met1-Ala232 Accession # Q8BJQ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353376
Test Specificity Detects mouse TRAM/TICAM2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TRAM/TICAM2 or rhTRIF/TICAM1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.