Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TRANCE/TNFSF11/RANK L Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB462SP
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Mouse samples. It is validated for Western Blot.Specifications
TRANCE/TNFSF11/RANK L | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8600 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
88227 | |
Western Blot 1 ug/mL | |
O35235 | |
TNFSF11 | |
Mouse myeloma cell line NS0-derived recombinant mouse TRANCE Arg72-Asp316 Accession # O35235 | |
25 μg | |
Primary | |
Detects mouse TRANCE in direct ELISAs and Western blots. In direct ELISAs, shows 100% cross-reactivity with recombinant human (rh) TRANCE and no cross-reactivity with rhAPRIL, rhBAFF, rhCD27 Ligand, recombinant mouse (rm) CD27 Ligand, rhCD30 Ligand, rmCD30 Ligand, rhCD40 Ligand, rmEDA, rhFas Ligand, rmFas Ligand, rhGITR Ligand, rhTNF-alpha , rhTRAK, rmTSG, recombinant cotton rat TNF-alpha , rmTNF-alpha , recombinant porcine TNF-alpha , recombinant rat TNF-alpha , and rhVEGI. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction