Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TRANCE/TNFSF11/RANK L Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Supplier: R&D Systems AF462
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
TRANCE/TNFSF11/RANK L | |
Polyclonal | |
Unconjugated | |
O35235 | |
TNFSF11 | |
Mouse myeloma cell line NS0-derived recombinant mouse TRANCE/TNFSF11/RANK L Arg72-Asp316 Accession # O35235 | |
100 μg | |
Primary | |
Detects mouse TRANCE/TNFSF11/RANK L in ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) CD27, rmCD40, and rmFas is observed and less than 1% cross-reactivity with rmCD30, recombinant human (rh) DR6, rmGITR, rmTNF RI, and rmTNF RII is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Goat | |
Affinity Purified | |
RUO | |
8600 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction