Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TRANCE/TNFSF11/RANK L Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems BAF462
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Polyclonal specifically detects TRANCE/TNFSF11/RANK L in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
| TRANCE/TNFSF11/RANK L | |
| Polyclonal | |
| Biotin | |
| O35235 | |
| TNFSF11 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TRANCE (R&D Systems, Catalog # 462-TR) Arg72-Asp316 Accession # O35235 | |
| 50 μg | |
| Primary | |
| Detects mouse TRANCE in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) TRANCE, recombinant mouse (rm) RANK, rhCD40 Ligand, and rmOPG is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8600 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction