Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse/Rat TREM-1 Quantikine ELISA Kit

Catalog No. MTRM10
Click to view available options
Quantity:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Mouse/Rat TREM-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat TREM-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat TREM-1 in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 31.30 - 2000pg/mL
Assay Sensitivity 5.4pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and Recombinant Mouse and Rat TREM-1
For Use With (Application) ELISA
Gene ID (Entrez) 58217
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Tissue Homogenates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD354, CD354 antigen, TREM1, TREM-1 Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Target TREM1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.