Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TREM2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1729SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1729SP 25 μg
MAB1729100 100 μg
MAB1729500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1729SP Supplier R&D Systems Supplier No. MAB1729SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

TREM2 Monoclonal specifically detects TREM2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TREM2
Applications Western Blot
Classification Monoclonal
Clone 237916
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99NH8
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Gene Symbols TREM2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM-2 Leu19-Pro168 Accession # Q99NH8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54209
Test Specificity Detects mouse TREM-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) TREM-1, recombinant human TREM-2, or rmTREM-3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.