Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TREML1/TLT-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | TREML1/TLT-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TREML1/TLT-1 Polyclonal specifically detects TREML1/TLT-1 in Mouse samples. It is validated for Western Blot.Specifications
TREML1/TLT-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
TREML1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_082039 | |
340205 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREML1/TLT-1 Gly18-Cys178 Accession # NP_082039 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse TREML1/TLT-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human TREML1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title