Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TREML1/TLT-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2424
This item is not returnable.
View return policy
Description
TREML1/TLT-1 Polyclonal specifically detects TREML1/TLT-1 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
TREML1/TLT-1 | |
Polyclonal | |
Biotin | |
NP_082039 | |
TREML1 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREML1/TLT-1 Gly18-Cys178 Accession # NP_082039 | |
50 μg | |
Primary | |
Detects mouse TREML1/TLT-1 in ELISAs and Western blots. In sandwich ELISAs, less than 0.03% cross-reactivity with recombinant human (rh) TREML1/TLT-1 and rhTREML2/TLT-2 is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
dJ238O23.3, GLTL1825, PRO3438, TLT1, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
340205 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction