Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TREML2/TLT-2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6707V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6707V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6707V100 Supplier R&D Systems Supplier No. FAB6707V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TREML2/TLT-2 Monoclonal specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TREML2/TLT-2
Applications Flow Cytometry
Classification Monoclonal
Clone 656906
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q2LA85
Gene Alias C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473
Gene Symbols TREML2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270 Accession # Q2LA85
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79865
Test Specificity Detects mouse TREML2/TLT-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) TREM-1, -2, -2b, -3, -4, rmTREML1/TLT-1, rmTREML4/TLT-4, or recombinant human (rh) NKp44 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.