Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TREML2/TLT-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB6707SP
This item is not returnable.
View return policy
Description
TREML2/TLT-2 Monoclonal specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
TREML2/TLT-2 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q2LA85 | |
TREML2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2 His25-Ala270 Accession # Q2LA85 | |
25 μg | |
Primary | |
Detects mouse TREML2/TLT-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) TREM-1, -2, -2b, -3, -4, rmTREML1/TLT-1, rmTREML4/TLT-4, or recombinant human (rh) NKp44 is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
656906 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
79865 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction