Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TRIM5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3550
This item is not returnable.
View return policy
Description
TRIM5 Polyclonal specifically detects TRIM5 in Mouse samples. It is validated for Western Blot.Specifications
TRIM5 | |
Polyclonal | |
Unconjugated | |
AAH38937 | |
TRIM5 | |
E. coli-derived recombinant mouse TRIM5 Ala2-Gln249 Accession # AAH38937 | |
100 μg | |
Primary | |
Detects mouse TRIM5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TRIM5 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
EC 6.3.2, EC 6.3.2.-, RING finger protein 88, RNF88, RNF88tripartite motif protein TRIM, TRIM5alpha, tripartite motif containing 5, tripartite motif protein TRIM5, tripartite motif-containing 5, tripartite motif-containing protein 5 | |
Goat | |
Affinity Purified | |
RUO | |
85363 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction