Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TrkC Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1404
This item is not returnable.
View return policy
Description
TrkC Polyclonal specifically detects TrkC in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TrkC | |
Polyclonal | |
Biotin | |
Q6VNS1 | |
NTRK3 | |
Mouse myeloma cell line NS0-derived recombinant mouse TrkC Cys32-Thr429 Accession # Q6VNS1 | |
50 μg | |
Primary | |
Detects mouse TrkC in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) TrkC is observed and less than 2% cross-reactivity with recombinant mouse TrkB, recombinant rat TrkA, and rhTrkA is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, NTRK3, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
Goat | |
Affinity Purified | |
RUO | |
4916 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction