Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TROP-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | TROP-2 |
---|---|
Clone | 168215 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
TROP-2 Monoclonal specifically detects TROP-2 in Mouse samples. It is validated for Western Blot.Specifications
TROP-2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q8BGV3 | |
4070 | |
Mouse myeloma cell line NS0-derived recombinant mouse TROP-2 Ser82-Thr268 Accession # Q8BGV3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TROP-2 in direct ELISAs and Western blots. |
168215 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, gp50, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, T16, TACSTD2, TROP2, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
TACSTD2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title