Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TROY/TNFRSF19 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB723
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for Western Blot.Specifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9JLL3 | |
| TNFRSF19 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TROY/TNFRSF19 Glu30-Leu170 Accession # Q9JLL3 | |
| 100 μg | |
| Primary | |
| Detects mouse TROY/TNFRSF19 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) 4-1BB, rmBAFF R, recombinant human RELT, rmCD3, rhDR6, rmDR3, rhTRADD, rmEDAR, recombinant rat FAS, rmFAS, rmGITR, rmHVEM, rmLymphotoxin beta R, rmNGF R, rmOPG, rmRANK, rmTNF RII, rmDcTRAIL R2, rmDcTRAIL RI, rmOX40, rhTRAIL R3, rhTNF RII, rmTWEAK, rmTRAIL, rmCD27, rmCD40, rhTNF RI, or rmTNF RI is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 323606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55504 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction