Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse TROY/TNFRSF19 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF723
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF723 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF723 Supplier R&D Systems Supplier No. BAF723
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TROY/TNFRSF19 Polyclonal specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen TROY/TNFRSF19
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q9JLL3
Gene Alias TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Gene Symbols TNFRSF19
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TROY/TNFRSF19 Glu30-Leu170 Accession # Q9JLL3
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55504
Test Specificity Detects mouse TROY/TNFRSF19 in ELISAs and Western blots. In sandwich immunoassays, less than 2% cross-reactivity with recombinant human (rh) TAJ is observed and less than 0.2% cross-reactivity with recombinant mouse (rm) EDAR, rmCD27, rmFas, rmRANK, rmTNF RI, and rmTNF RII is observed. In Western blots, approximately 5% cross-reactivity with rmCD27 and rmEDAR is observed and less than 1% cross-reactivity with rhTNF RI, rhTNF RII, rmOPG, rmRANK, rmCD30, rmCD40, rm4-1BB, rmFas, and rmGITR (under non-reducing conditions) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.