Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Mouse TROY/TNFRSF19 ELISA Kit (Colorimetric)
SDP

Catalog No. NB228873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB228873 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB228873 Supplier Novus Biologicals Supplier No. NBP280369

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 7.88 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Mouse TROY/TNFRSF19.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 55504
Gene Symbol TNFRSF19
Quantity 1 Kit
Research Discipline Apoptosis
Storage Requirements Storage is content dependent.
Synonym TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.